Learn More
Novus Biologicals™ Purple Hematoxylin Solution
Brand:  Novus Biologicals™ NBP3-12176
Product Code. 18689510
  • 470.00€ / 100mL
Explore more special offers
Specifications
Store at room temperature.
Immunohistochemistry, Immunohistochemistry-Paraffin
100 mL
Preparation of Working Solutions Purple Hematoxylin is ready to use and does not require any preparation. Protocol Recommendations Counterstain 1. Rinse slides with deionized or distilled water. 2. Place slides into Purple Hematoxylin for 30-60 seconds. 3. Rinse slides in deionized or distilled water until all free hematoxylin has been removed. 4. Bluing the slides in an alkaline solution is not required and is not recommended.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Novus Biologicals™ Purple Hematoxylin Solution > 100 mL