Learn More
Euro Protection™ Purofort Safety Boot
86.47€
Specifications
Certifications/Compliance | EN 345:1992 CLASSE S5.Ci, CE |
---|---|
Design | Slim |
For Use With (Application) | Industries, Handling heavy objects, cold environments or winter, Agriculture, slippery Media, Oil Media, Wetlands or wet weather |
Grade | S5 |
Liner Material | Polyamide |
Specifications
EN 345:1992 CLASSE S5.Ci, CE | |
Industries, Handling heavy objects, cold environments or winter, Agriculture, slippery Media, Oil Media, Wetlands or wet weather | |
Polyamide | |
Polyurethane |
Slim | |
S5 | |
Polyurethane, Polyamide | |
Purofort Safety Boot |