Learn More
Keystone Cleanroom Products PureClean Self-Sealing 1421B Tyvek™ Autoclave Bag with Clear Film Side and Steam Indicator, 10 to 15 in. wide
GREENER_CHOICE
PureClean™ Self-Sealing Autoclave Bags are made from a transparent film and 1421B DuPont™ Tyvek™, which is a new grade of uncoated, untreated, flexible Tyvek™ material specifically designed for pharmaceutical use in steam sterilization applications.
3295.00€ - 5270.00€
Specifications
Product Type Autoclave Bag
Autoclavable Autoclavable
Certifications/Compliance COC
Closure Type Safe Seal
Color Clear, White
View More Specs
Products 38
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
30042304
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
6STKRP1424SS
14 x 24
4210.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
30042225
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
6STKRP1420SS
14 x 20
4165.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
30042317
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
6STKRP1418SS
14 x 18
4070.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
30042243
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
6STKRP1416SS
14 x 16
4045.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
30042278
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
6STKRP1414SS
14 x 14
3955.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
30042299
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
6STKRP1248SS
12 x 48
4165.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
30042280
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
6STKRP1246SS
12 x 46
4070.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
30042203
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
6STKRP1242SS
12 x 42
4000.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
30042179
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
6STKRP1240SS
12 x 40
3955.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042244
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1236SS
12 x 36
3860.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042321
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1234SS
12 x 34
3745.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042207
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1232SS
12 x 32
3690.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042205
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1230SS
12 x 30
3645.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042277
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1228SS
12 x 28
3600.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042315
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1224SS
12 x 24
3555.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042231
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1222SS
12 x 22
3365.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042188
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1220SS
12 x 20
3365.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042189
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1218SS
12 x 18
3365.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042257
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1216SS
12 x 16
3345.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042255
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1214SS
12 x 14
3320.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042216
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1212SS
12 x 12
3295.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042248
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1062SS
10 x 62
5010.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042191
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1056SS
10 x 56
5010.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042251
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1048SS
10 x 48
4915.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042270
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1036SS
10 x 36
3860.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042220
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1030SS
10 x 30
4255.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042228
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1024SS
10 x 24
3510.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042212
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1022SS
10 x 22
3390.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042246
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1020SS
10 x 20
3345.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042184
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1018SS
10 x 18
3345.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042206
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1015SS
10 x 15
3295.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042201
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1543SS
15 x 43
4775.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042263
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1526SS
15 x 26
4210.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042208
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1448SS
14 x 48
5270.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Aggiungi al carrello
30042302
GREENER_CHOICE
Vedere i documenti
Keystone Cleanroom Products
6STKRP1440SS
14 x 40
4565.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
30042281
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
6STKRP1438SS
14 x 38
4490.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
30042215
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
6STKRP1436SS
14 x 36
4400.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
30042193
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
6STKRP1426SS
14 x 26
4355.00€
Pack of 250
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
Description
Description
  • Uncoated Tyvek™ eliminates potential of residue deposits on equipment, post sterilization
  • Clear Side enables visual confirmation of dryness for sterilized parts and equipment, which is required by EU GMP Annex 1
  • Convenient Self-Seal Closure eliminates the need for autoclave tape or the validation and use of a heat sealer
  • Fast and consistent for equipment preparation area
  • Resists tearing and punctures
  • Superior microbial barrier
  • Manufactured and packaged in a certified ISO 6 cleanroom
  • Made in the USA
  • Certificates of Conformance sent with shipment
Specifications
Specifications
Autoclave Bag
COC
Clear, White
Steam Sterilization of Pharmaceutical Equipment
Non-sterile
Yes
5.3 mil
Autoclavable
Safe Seal
Single-use
Tyvek
5.3 mil
121 to 125 C
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Keystone Cleanroom Products PureClean Self-Sealing 1421B Tyvek™ Autoclave Bag with Clear Film Side and Steam Indicator, 10 to 15 in. wide