Learn More
Quickfit™ PTFE Joint Clips with Conical Joints
Highly resistant to chemical attack
Brand:  Quickfit GC12
34.11 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12009062
  • 36.80€ / Pack of 5
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Description
Highly resistant to chemical attack
  • Used for retaining glassware having Quickfit™ conical joints
  • Manufactured from PTFE with encapsulated steel spring to provide firm grip
  • Clips are numbered with joint size for easy identification
  • Can be used at temperatures up to 250°C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Joint Clip
Encapsulated Steel spring
12/21
PTFE
Conical Joints
250°C