Learn More
PST1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-41063JF646
Product Code. 18748532
  • 589.00€ / 0.10mL
Explore more special offers
Description
Description
PST1 Polyclonal antibody specifically detects PST1 in Yeast samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PST1
Polyclonal
Western Blot, ELISA
Cell wall mannoprotein PST1, D4214, Haze protective factor 2, HPF2, YD9609.09, YDR055W
Antibody was raised against a synthetic peptide from the amino terminus of yeast Pst1. The immunogen is located within amino acids 80 - 130 of Pst1. Amino Acid Squence: DSLESITDSLNLQSLT
0.1 mL
Infections (Virus Bacteria and Parasites)
851625
Store at 4C in the dark.
IgG
Western Blot, ELISA, Immunohistochemistry (Paraffin)
Janelia Fluor 646
50mM Sodium Borate
Rabbit
Peptide affinity purified
RUO
Primary
Yeast
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
PST1 Antibody, Janelia Fluor™ 646, Novus Biologicals™ > 0.1 mL; Janelia Fluor 646