missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PPAR gamma/NR1C3 Recombinant Protein Antigen

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

Brand:  Novus Biologicals™ NBP2-57833PEP

Product Code. 18274113

  • 259.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human PPAR gamma/NR1C3. Source: E.coli Amino Acid Sequence: DYKYDLKLQEYQSAIKVEPASPPYYSEKTQLYNKPHEEPSNSLMAIECRVC The PPAR gamma/NR1C3 Recombinant Protein Antigen is derived from E. coli. The PPAR gamma/NR1C3 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5468
PPAR gamma/NR1C3 Recombinant Protein Antigen
PBS and 1M Urea, pH 7.4
CIMT1, NR1C3GLM1, Nuclear receptor subfamily 1 group C member 3, peroxisome proliferator-activated receptor gamma, peroxisome proliferator-activated receptor gamma 1, PPAR gamma, PPARG1peroxisome proliferative activated receptor, gamma, PPARG2PPARgamma, P
Unlabeled
100 μL
E.coli
>80% by SDS-PAGE and Coomassie blue staining
Store at −20°C. Avoid freeze-thaw cycles
Blocking/Neutralizing, Control
PPARG
Recombinant Protein Antigen
RUO
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-50015. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ PPAR gamma/NR1C3 Recombinant Protein Antigen > Quantity: 100μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For research use only.