Learn More
Labconco Power Cord BS-1363 13A/250V (UK)
Power Cord BS-1363 13A/250V (UK) is a replacement UK power cord for 230V/50Hz Labconco freeze dryer models.
Brand:  Labconco 1332601
35.09 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 18085461
  • 36.00€ / Each
Estimated Shipment: 10-02-2026
to see stock.
Explore more special offers
Description
  • Power Cord BS-1363 is 13A 250 V
  • The plug type is UK
  • Serves as a replacement UK power cord for 230 V/50 Hz Labconco freeze dryer models
Specifications
Specifications
Power Cord
250 V
Documents
One moment while we fetch your results.
Product Title
Labconco Power Cord BS-1363 13A/250V (UK) > Power cord: Amps: 13; Voltage: 250; Plug Type: UK