Learn More
Invitrogen™ Power Blotter XL System
Get rapid and efficient semi-dry protein transfer with the Invitrogen™ Power Blotter.
Brand:  Invitrogen™ PB0013
Product Code. 15889061
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Includes: Invitrogen Power Blotter XL Cassette, Invitrogen Power Blotter Station
Description
Description
The Invitrogen™ Power Blotter System is specifically designed for rapid semi-dry transfer of proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes in 5 to 12 minutes. The Power Blotter includes an integrated power supply optimized to enable consistent and high-efficiency protein transfer.
The Power Blotter Cassette enables the simultaneous transfer of gels. The Power Blotter Station has an easy-to-use color LCD touchscreen interface with pre-programmed transfer methods for different numbers and sizes of gels and ranges of molecular weight proteins. The easy-touch programming feature allows custom transfer settings to be quickly created, saved, and run.
The Power Blotter achieves highly efficient transfer in 7 minutes when used with Power Blotter Select Transfer Stacks or 10 minutes with the Power Blotter 1-Step Transfer Buffer. The instrument is also effective for standard 30- to 60-minute semi-dry transfer protocols based on traditional Towbin buffer.
Features:
  • Efficient—high transfer efficiency with a broad range of protein sizes compared to conventional semi-dry or wet (tank) transfer methods
  • Fast—5 to 12 minute transfer when used with Power Blotter Select Transfer Stacks or Power Blotter 1-Step Transfer Buffer
  • Integrated power supply—seamless operation between control unit and cassette; no need for external power supply
  • Easy-touch programming—convenient pre-programmed transfer methods or create, save, and run customized transfer methods
  • Flexible—simultaneous transfer
  • Versatile—compatible with Power Blotter Select Transfer Stacks, Pierce 1-Step Transfer Buffer for rapid blotting programs or Towbin transfer buffer for conventional semi-dry transfer methods
TRUSTED_SUSTAINABILITY
Specifications
Specifications
• Power Blotter Station
• Power Blotter Cassette XL
Nitrocellulose, PVDF
Power Blotter
Blotter System
Room Temperature
Western Blot
Semi-Dry
2 Midi or 4 Mini Gels
1 system