missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
Boiling Point | 1320°C (1013 hPa) |
CAS | 1310-58-3 |
Melting Point | 360°C |
Assay Percent Range | min. 86.0% |
Molecular Formula | HKO |
MDL Number | MFCD00003553 |
Density | 2.04 g/cm³ (20°C) |
Synonym | Caustic potash |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Quantity | 1 kg |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling


Signal Word
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium hydroxide, BAKER ANALYZED™ ACS, J.T. Baker™ >
Spot an opportunity for improvement?Share a Content Correction