Learn More
Brand™ Polystyrene Semi-Micro Cuvette
Constructed of polystyrene and acrylic glass. BRAND™ Polystyrene Semi-Micro Cuvettes provide a clear, clean optical path with indication of optical path orientation.
Brand: Brand™ 759015
14.48 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Constructed of polystyrene and acrylic glass.
- Clear, clean optical path with indication of optical path orientation
- Manufactured under controlled room conditions and packaged fully automatically, without human contact
- Packed grouped by mold cavity number to ensure lowest variation of extinction coefficient
Specifications
1.5 mL | |
4.5mm | |
360 nm |
Polystyrene | |
100 Pack | |
23 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
0000013340 | Certificate of Analysis | 07/02/2025 | 759015 |
0000013180 | Certificate of Analysis | 20/01/2025 | 759015 |
0000012998 | Certificate of Analysis | 25/11/2024 | 759015 |
0000012586 | Certificate of Analysis | 07/10/2024 | 759015 |
0000012557 | Certificate of Analysis | 07/10/2024 | 759015 |
Product Content Correction
Product Title
Brand™ Polystyrene Semi-Micro Cuvette > Volume: 1.5mL