Learn More
BRAND™ Polypropylene Thread Adapters
These Polypropylene (PP) and Ethylene Tetrafluoroethylene (ETFE) adapters are for general purpose use and are ideal for use with Dispensetter bottletop dispensers.
Brand: BRAND™ 704325
6.45 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Thread adapter, Polypropylene (PP), outer thread GL 45, for bottle thread S 40
- Thread Adapters for BRAND™ Bottle Top Dispensers, Polypropylene, 45/40 mm
- For use with Bottle Thread 40 S, Dispensette, Seripettor, Titrette and QuikSip Bottletop Diluters and Dispensers
- The Polypropylene (PP) and Ethylene Tetrafluoroethylene (ETFE) adapters are for general purpose use
- Ideal for use with Dispensetter bottletop dispensers
- These polypropylene and fluoropolymer (ETFE and PTFE) adapters are for use with BRAND™ bottletop dispensers and burettes
- These thread adapters help to maximize the compatibility of bottletop dispensers with most reagent bottles
Specifications
Thread Adapter | |
For bottle thread GL 24-25 | |
32 GL |
Polypropylene | |
1 Ea. |
Product Content Correction
Product Title
BRAND™ Polypropylene Thread Adapters > For Use With: Bottle Thread 24 to 25 GL, Dispensette, Seripettor, Titrette and QuikSip