Learn More
Gosselin™ Polypropylene Straight Specimen Containers
CORNING GOSSELIN™ Straight Container, Specimen Vials, PP come in different sizes and quantities, and are available sterile and non-sterile.
Brand: Gosselin™ TP30C-030
This item is not returnable.
View return policy
Description
- Clear and break-resistant polypropylene (PP)
- Available in different sizes and quantities
- Color-coded screw caps
Specifications
Specimen Container | |
Polypropylene | |
Non-sterile | |
Clear | |
1200/Cs. |
No | |
Straight | |
40 mL | |
No Label | |
1200/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Gosselin™ Polypropylene Straight Specimen Containers > 40mL