Learn More
Fisherbrand™ Polypropylene Dispenser Tips: Adapter
Polypropylene made adapter for dispenser tip 25/50 mL that is ideal in most research, scientific, and laboratory environments.
Brand:  Fisherbrand™ 702640
 View more versions of this product
Product Code. 15103666
  • 37.30€ / Pack of 5
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Adapter for dispenser tips
  • 25/50mL
  • Sterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sterile
SDS
Documents Certificates
Promotions
Promotions
  • Fisherbrand
    Get up to 60% off select Fisherbrand products
    Promo Code: 24163
    Expires2025-04-30
    Days
    7
    Hours
    9
    Minutes
    4
    Seconds
    25
    New Offer
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/04/2025 and 30/04/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions Save Now
Product Content Correction
Product Title
Fisherbrand™ Polypropylene Dispenser Tips: Adapter >