missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Polypropylene 96-well F-Bottom Microplates

Manufactured from polypropylene for high resistance to common chemicals and thermal stability. Greiner Bio-One–Polypropylene 96 Well Microplates– are ideal for storage of active reagents, patient samples or biomolecules.

Brand:  Greiner Bio-One 655207

 View more versions of this product

Product Code. 10764114

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Traceability to production through a defined LOT number system
  • Applications: - Long-term storage of active agents - Storage of patient samples in diagnostics - Storage of DNA or RNA, stock cultures
  • Uniform external dimensions
  • Well-to-well spacing 9 mm
  • Alphanumeric well coding High chemical resistance
  • High temperature resistance (-196 to +121°C)
  • Sealable with adhesive films and heat sealer or CapMats
  • Solid bottom
  • Well profile: F-Bottom/chimney well (flat bottom)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
White
96
Flat Bottom (F-well)
–196 to 121°C
8 x 12
Polypropylene
Non-sterile
Long-term storage of active agents
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.