missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Polypropylene 96-well F-Bottom Microplates
Manufactured from polypropylene for high resistance to common chemicals and thermal stability. Greiner Bio-One–Polypropylene 96 Well Microplates– are ideal for storage of active reagents, patient samples or biomolecules.
332.94€ - 480.00€
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Material | Polypropylene |
No. of Wells | 96 |
Well Shape | Flat Bottom (F-well) |
Product Code | Brand | Color | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Sterility | Price | Quantity & Availability | ||||
10093241
|
Greiner Bio-One
655261 |
Clear | Sterile |
389.00€
Pack of 100 |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||||
10131992
|
Greiner Bio-One
655201 |
Clear | Non-sterile |
371.00€
Pack of 100 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
||||
10706172
|
Greiner Bio-One
655209 |
Black | Non-sterile |
480.00€
Pack of 100 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
||||
10764114
|
Greiner Bio-One
655207 |
White | Non-sterile | N/A | |||||
Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Traceability to production through a defined LOT number system
- Applications: - Long-term storage of active agents - Storage of patient samples in diagnostics - Storage of DNA or RNA, stock cultures
- Uniform external dimensions
- Well-to-well spacing 9 mm
- Volume–Total volume: 392μL; Working volume: 25 to 370μL
- Alphanumeric well coding
- Sealable with adhesive films and heat sealer or CapMats
- Solid bottom
Specifications
Standard | |
Polypropylene | |
Flat Bottom (F-well) |
8 x 12 | |
96 | |
Long-term storage of active agents |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title