missing translation for 'onlineSavingsMsg'
Learn More
Poly(ethylene glycol), average M.W. 4000, Thermo Scientific Chemicals Product Code.: 10169679
Add to basket

Poly(ethylene glycol), average M.W. 4000, Thermo Scientific Chemicals

Product Code. 10169679
1 kg, Plastic Bottle
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10169679

Brand: Thermo Scientific Acros 434630010

Add to basket
Add to basket
Request Bulk or Custom Format
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEO
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color White
Physical Form Flakes
Melting Point 54.0°C to 58.0°C
Linear Formula H(OCH2CH2)nOH
Merck Index 15,7688
Quantity 1 kg
Formula Weight 4000
Infrared Spectrum Authentic
Packaging Plastic Bottle
Water 0.5% max.
Chemical Name or Material Poly(ethylene glycol)
Show More Show Less
Product Identifier
  • Poly(ethylene glycol), average M.W. 4000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Poly(ethylene glycol), average M.W. 4000, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.