Learn More
Thermo Scientific™ Plate Count Agar (Bottled)
Enumerate viable organisms in milk, dairy products, water, food and other specimens of sanitary importance using these ready-prepared media bottles.
158.00€
Specifications
Color | Straw, Clear |
---|---|
Content And Storage | 2 to 25°C |
Format | Sirop Bottle |
Manufacturing Origin | United Kingdom |
Product Type | Agar |
Product Code | Brand | Fill Weight | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Fill Weight | Price | Quantity & Availability | |||||
12643408
|
Thermo Scientific™
BO0195V |
500.0g + 4.0g |
158.00€
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Enumerate viable organisms in milk, dairy products, water, food and other specimens of sanitary importance using non-selective Thermo Scientific™ Oxoid™ Plate Count Agar ready-prepared bottled medium. Plate Count Agar is equivalent to the medium recommended by the APHA1, PHLS 2 and ISO3,4 for the plate count of microorganisms in food.
Remove the need for autoclaving and dispensing with convenient, pre-filled and ready-to-use bottles of Plate Count Agar.
- Convenient: pre-filled bottles are ready for immediate use.
- Simple: easy-to-use with minimum training.
- Long shelf life: upto 12 months.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
Straw, Clear | |
Sirop Bottle | |
Agar | |
10/Pk. |
2 to 25°C | |
United Kingdom | |
10 bottles with screw cap closures in a box |
Safety and Handling
missing translation for 'shelfLife' : 52 Weeks
Your input is important to us. Please complete this form to provide feedback related to the content on this product.