Learn More
Fisherbrand™ Polystyrene Petri Dishes
Brand:  Fisherbrand™ FB63186
67.36 EUR valid until 2025-12-31
Use promo code "24345" to get your promotional price.
Product Code. 12694785
  • 146.00€ / Pack of 600
In Stock
to see stock.
Edge
Explore more special offers
Specifications
Polystyrene
Low Form Petri Dish
3
90 mm
Non-sterile
600 Case
14.2 mm
Videos
  • Fisherbrand - Advantage aspiration system
    The Fisherbrand aspiration Advantage system is a compact and portable unit with an integrated resistant diaphragm pump, used for the safe and precise aspiration of non-flammable chemical, biological, and medical liquids.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Fisherbrand Advantage aspiration system
    SDS
    Documents Certificates
    Certificates
    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
    4 results found
    Lot Number Certificate Type Date Catalog Number
    Lot NumberM554720 Certificate TypeCertificate of Compliance Date11/03/2025 Catalog NumberFB63186
    Lot NumberM553990 Certificate TypeCertificate of Compliance Date28/02/2025 Catalog NumberFB63186
    Lot NumberM553230 Certificate TypeCertificate of Compliance Date06/02/2025 Catalog NumberFB63186
    Lot NumberM551990 Certificate TypeCertificate of Compliance Date29/01/2025 Catalog NumberFB63186
    Product Title
    Fisherbrand™ Polystyrene Petri Dishes > Triple Vented