Learn More
Gilson™ PIPETMAN G 4-Pipette Kit
PIPETMAN G builds a new level of comfort into the Gilson™ pipette range with a redesigned piston that significantly reduces the force needed to aspirate and dispense liquids, a benefit during extended pipetting sessions which helps alleviate common hand stresses.
Brand: Gilson™ F167360
1091.12 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Contains: 4 PIPETMAN G Pipettes (P2G, P20G, P200G, and P1000G), 3 racks of certified quality PIPETMAN DIAMOND Tips (D10L, D200 and D1000), 4 SINGLE™ pipette holders, and 1 Gilson Guide to Pipetting
- Maintains the trusted durability, accuracy and reliability of the PIPETMAN Classic
- Requires some of the lowest pipetting forces to operate (3 newetons on average)
- Lifetime warranty
- Manual air displacement type
Specifications
Polypropylene | |
1 | |
Manual Air Displacement Pipette Kit |
Pipetman G | |
Pipetman G | |
0.2-2 μL, 100-1000 μL, 2-20 μL, 20-200 μL |
Promotions
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%