missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperacillin Sodium Salt, MP Biomedicals™
117.66€ - 800.30€
Chemical Identifiers
CAS | 59703-84-3 |
---|---|
Molecular Formula | C23H26N5NaO7S |
Molecular Weight (g/mol) | 539.54 |
MDL Number | MFCD00917471 |
InChI Key | WCMIIGXFCMNQDS-QMJSIVKPSA-M |
Synonym | Sodium Piperacillin, Pipracil |
PubChem CID | 23666879 |
ChEBI | CHEBI:8233 |
SMILES | [Na+].CCN1CCN(C(=O)N[C@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C2=CC=CC=C2)C(=O)C1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11413652
|
MP Biomedicals
0215626801 |
1 g |
117.66€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
11433652
|
MP Biomedicals
0215626810 |
10 g |
800.30€
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
59703-84-3 | |
539.54 | |
WCMIIGXFCMNQDS-QMJSIVKPSA-M | |
23666879 | |
[Na+].CCN1CCN(C(=O)N[C@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C2=CC=CC=C2)C(=O)C1=O |
C23H26N5NaO7S | |
MFCD00917471 | |
Sodium Piperacillin, Pipracil | |
CHEBI:8233 |
Specifications
Piperacillin Sodium Salt | |
59703-84-3 | |
MFCD00917471 | |
WCMIIGXFCMNQDS-QMJSIVKPSA-M | |
sodium;(2S,5R,6R)-6-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate | |
23666879 | |
539.5 | |
Piperacillin Sodium Salt |
Crystalline Powder | |
C23H26N5NaO7S | |
Sodium Piperacillin, Pipracil | |
[Na+].CCN1CCN(C(=O)N[C@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C2=CC=CC=C2)C(=O)C1=O | |
539.54 | |
CHEBI:8233 | |
White |
Safety and Handling
Recommended Storage : Store at 4°C.
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- piperacillin sodium salt
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P321-Specific treatment (see on this label).
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : pipperacillin sodiumsalt