missing translation for 'onlineSavingsMsg'
Learn More

Piperacillin Sodium Salt, MP Biomedicals™

Product Code. 11413652
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 59703-84-3
Molecular Formula C23H26N5NaO7S
Molecular Weight (g/mol) 539.54
MDL Number MFCD00917471
InChI Key WCMIIGXFCMNQDS-QMJSIVKPSA-M
Synonym Sodium Piperacillin, Pipracil
PubChem CID 23666879
ChEBI CHEBI:8233
SMILES [Na+].CCN1CCN(C(=O)N[C@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C2=CC=CC=C2)C(=O)C1=O

Specifications

Type Piperacillin Sodium Salt
Physical Form Crystalline Powder
IUPAC Name sodium;(2S,5R,6R)-6-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Formula Weight 539.5
Color White
Quantity 1 g
Chemical Name or Material Piperacillin Sodium Salt
compliance-icons compliance-icons
Product Identifier
  • piperacillin sodium salt
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P321-Specific treatment (see on this label).
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : pipperacillin sodiumsalt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'