missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pinometostat, 95%

Brand:  Thermo Scientific™ 468161000

553.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 17141136

  • 555.77€ / 100mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pinometostat
EPZ-5676
C30 H42 N8 O3
Solid
95%
1380288-87-8
100 mg
562.71
Reagent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number344427 Certificate TypeCertificate of Analysis Date19/10/2020 Catalog Number
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pinometostat, 95% > 100 mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.