missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Phosphate Test Kits
Obtain precision measurements for waste water, drinking water, surface water, and fertilizers with CombiCheck Phosphate (PO4) cell test kits.
140.00€ - 180.00€
Specifications
For Use With (Application) | For waste water, drinking water, surface water, fertilizers |
---|---|
Includes | Certificates |
Sufficient For | 25 Tests |
Test Method | Photometric |
Product Code | Brand | Type | No. of Tests | Range | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | No. of Tests | Range | Price | Quantity & Availability | |||
11777315
|
WTW™
252076 |
CombiCheck Phosphate (PO4) cell test kit | 90 | 0.5 to 25 mg/L PO4-P, 0.5 to 25 mg/L Total, 1.5 to 76.7 mg/L PO4 |
144.00€
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11771093
|
WTW™
250413 |
CombiCheck Phosphate (PO4) cell test kit | 25 | 0.5 to 25 mg/L PO4-P, 1.5 to 76.7 mg/L PO4 |
140.00€
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10327481
|
WTW™
250446 |
CombiCheck COD Chemical oxygen demand O2 cell test kit | 95 | 0.010 to 5 mg/L PO4-P, 0.03 to 15.3 mg/L PO4 |
171.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10035410
|
WTW™
250447 |
n-Ten (nonionic) surfactant cell test kit | 25 | 0.5 to 30 mg/L PO4-P, 1.5 to 92 mg/L PO4 |
180.00€
2 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Variety of instrumentation for measuring phosphates
- PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
- For waste water, drinking water, surface water, fertilizers
- CombiCheck Phosphate (PO4) cell test kit
Specifications
For waste water, drinking water, surface water, fertilizers | |
Certificates | |
25 Tests | |
Photometric |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Phosphate Cell Test P7/25 Method: photometric 0.5 - 25.0 mg/l PO
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid