missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Phosphate Test Kits

Obtain precision measurements for waste water, drinking water, surface water, and fertilizers with CombiCheck Phosphate (PO4) cell test kits.

Brand:  WTW™ 250447


 View more versions of this product

Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10035410

  • 180.00€ / 2 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Variety of instrumentation for measuring phosphates
  • PhotoLab® S6, PhotoLab® S12, PhotoLab® Spectral, Photoflex photometers
  • For waste water, drinking water, surface water, fertilizers
  • CombiCheck Phosphate (PO<sub>4</sub>) cell test kit
TRUSTED_SUSTAINABILITY
Specifications

Specifications

For waste water, drinking water, surface water, fertilizers
Certificates
0.5 to 30 mg/L PO4-P, 1.5 to 92 mg/L PO4
Photometric
For PhotoLab™ S12, PhotoLab™ Spectral photometers
25
25 Tests
n-Ten (nonionic) surfactant cell test kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • phosphate test, o-phosphate

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, ammonium monovanadate

SDS
Documents

Documents

Certificates
Promotions

Promotions