Learn More
Savillex™ PFA Microtube Thread Caps
27.00€ - 129.00€
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | ||||||
11922165
|
Savillex™
60002401 |
13.7 mm |
27.00€
Each |
Estimated Shipment: 05-05-2025
Log in to see stock. |
||||||
11997495
|
Savillex™
60001183 |
2 mm |
129.00€
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||||
11962175
|
Savillex™
60003301 |
15.7 mm |
38.48€
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
Specifications
Microtube Thread Cap | |
PFA |
Product Title
Savillex™ PFA Microtube Thread Caps