missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Petri Dish Platinum Electrode for Tissues Chamber Kit

Petri Dish Platinum Electrode for Tissues Chamber Kit electrodes and chambers for ex vivo tissues gene or drug delivery.

1840.00€ - 2255.00€

Specifications

For Use With (Equipment) ECM™ 830, ECM™ 2001+, Gemini X2, Legacy ECM™ 2001
Material Platinum
Product Line BTX Electroporation
Product Type Electroporation accessory
Voltage 200 V
View More Specs

Products 2
Product Code Brand Size (Metric) Gap Price Quantity & Availability  
Product Code Brand Size (Metric) Gap Price Quantity & Availability  
15427340
View Documents
BTX™
450507
15 mm
2255.00€
Each
Please to purchase this item. Need a web account? Register with us today!
15407340
View Documents
BTX™
450505
5 mm
1840.00€
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Simple and easy to use
  • Available in 5 and 15 mm
  • Reusable, lab-grade Pyrex glass and two platinum electrodes
Specifications

Specifications

ECM™ 830, ECM™ 2001+, Gemini X2, Legacy ECM™ 2001
BTX Electroporation
200 V
10 μsec. to 100 msec.
Non-sterile
Square Wave
Platinum
Electroporation accessory
Petri Dish Electrode Chamber and Electrodes
1 Ea.
Banana Plug to Micrograbber
Videos
SDS
Documents

Documents

Product Certifications