missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Petri Dish Platinum Electrode for Tissues Chamber Kit

Petri Dish Platinum Electrode for Tissues Chamber Kit electrodes and chambers for ex vivo tissues gene or drug delivery.

Brand:  BTX™ 450507

 View more versions of this product

Product Code. 15427340

  • 2255.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Simple and easy to use
  • Available in 5 and 15 mm
  • Reusable, lab-grade Pyrex glass and two platinum electrodes
Specifications

Specifications

For ex Vivo Tissues Gene or Drug Delivery Using Large or Uniquely Shaped Tissue Samples
Platinum
Electroporation accessory
Petri Dish Platinum Electrode for Tissues Chamber Kit, 15 mm gap
5 mm
Petri Dish Electrode Chamber and Electrodes
10 mm
1 Ea.
15 mm
15 mm
Banana Plug to Micrograbber
ECM™ 830, ECM™ 2001+, Gemini X2, Legacy ECM™ 2001
BTX Electroporation
200 V
0.2 in.
Square
0.39 in.
10 μsec. to 100 msec.
0.59 in.
0.59 in.
Non-sterile
Square Wave
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions