missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentane, for Residue Analysis, Distol™, ≈95% N-Pentane
Click to view available options
Quantity:
1 L
2.5 L
Specifications
Specifications
Melting Point | -130°C |
Boiling Point | 36°C |
Quantity | 1 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 573mbar at 20°C |
Viscosity | 0.25 mPaS at 20°C |
Formula Weight | 72.15g/mol |
Safety and Handling




Product Identifier
- n-Pentane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pentane, for Residue Analysis, Distol™, ≈95% N-Pentane >
Spot an opportunity for improvement?Share a Content Correction