Learn More
3M™ PELTOR™ Optime™ II Ear Muffs
missing translation for 'orderingAttributeHoverText'
Style:
Headband
Passive Headset
Beschreibung
- Electrically insulated headband (dielectric) version available
- Large space inside cup helps reduce moisture and heat build-up
- Sealing rings filled with a unique combination of liquid and foam
- Easy-to-replace cushions and inserts helps keep them hygienically clean
- Soft, wide cusions help reduce pressure around the ears and improves comfort and wearability
Spezifikation
Spezifikation
Color | Green |
Certifications/Compliance | CE Approved |
For Use With (Application) | Suitable Choice for Noisy Industrial Environments for the Agricultural Sector, Airports or for the Use of Construction Equipment |
Plug Shape | Oval |
Product Type | Hearing Protection Optime 2 |
Material | Stainless Steel |
Style | Passive Headset |
Dokumentation
Product Information
Datasheets
7000039619
File size: 347.8 kB
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
3M™ PELTOR™ Optime™ II Ear Muffs >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur