Learn More
Herka™ Pear-shaped Tube
These pear-shaped, borosilicate glass centrifuge tubes are used in applications for the determination of water or sediment content in petroleum products. Graduated.
Brand: Herka™ A1129500
This item is not returnable.
View return policy
Description
- Graduated
Specifications
100 mL | |
203 mm | |
37 mm |
Borosilicate Glass | |
3.4 oz. | |
Yes |
Product Content Correction
Product Title
Herka™ Pear-shaped Tube > Pear-shaped Tube; Capacity: 100 mL