Learn More
Fisherbrand™ PE Plug for Shell Vial
Polyethylene caps are chemically inert and suitable for most chromatography applications.
Brand: Fisherbrand™ 15 08 0547
This item is not returnable.
View return policy
Description
- PE Plug
- Transparent
- Snap caps are easy to apply and easy to remove, eliminate the need for crimping or de-capping tools
Specifications
Transparent | |
Polyethylene | |
Plug |
15 mm | |
100 | |
ND15 |
Product Content Correction
Product Title
Fisherbrand™ PE Plug for Shell Vial > For Shell Vial 4ml,transparent