missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PCI 29732
Potent BTK inhibitor
281.00€ - 1100.00€
Specifications
Purity | >99% |
---|---|
Product Type | PCI 29732 |
Molecular Weight (g/mol) | 371.44 |
Inhibitors | PCI 29732 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18721799
|
Tocris Bioscience™
5012/10 |
10 mg |
281.00€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18773949
|
Tocris Bioscience™
5012/50 |
50 mg |
1100.00€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent BTK inhibitor (IC50 = 0.3 nM). Blocks B cell antigen receptor (BCR)-mediated gene expression in CD20+ B cells.Specifications
>99% | |
PCI 29732 | |
371.44 | |
PCI 29732 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title