Learn More
Tocris Bioscience™ PCI 29732
Potent BTK inhibitor
Brand:  Tocris Bioscience™ 5012/10
 View more versions of this product
Product Code. 18721799
  • 281.00€ / 10mg
Explore more special offers
Description
Description
PCI 29732 is a potent BTK inhibitor (IC50 = 0.3 nM). Blocks B cell antigen receptor (BCR)-mediated gene expression in CD20+ B cells.
Specifications
Specifications
>99%
PCI 29732
PCI 29732
10 mg
371.44