missing translation for 'onlineSavingsMsg'
Learn More
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals Product Code.: 10357830

p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals

Product Code. 10357830
2.5 kg, Plastic bottle
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10357830

Brand: Thermo Scientific Acros 171780025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6192-52-5
MDL Number MFCD00142137
InChI Key KJIFKLIQANRMOU-UHFFFAOYSA-N
Synonym p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
PubChem CID 521998
IUPAC Name 4-methylbenzenesulfonic acid;hydrate
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.O

Specifications

Melting Point 100.0°C to 108.0°C
Color Pink to White
Flash Point 180°C
Infrared Spectrum Authentic
Assay Percent Range 97.5%
Packaging Plastic bottle
Molecular Formula C7H8O3S·H2O
Linear Formula CH3C6H4SO3H·H2O
Quantity 2.5 kg
Beilstein 11, 97
Fieser 01,1172; 04,508; 05,673; 06,597; 07,374; 08,488; 09,471; 11,535; 12,507
Merck Index 15, 9692
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and chloroform
Molecular Weight (g/mol) 190.22
Formula Weight 190.22
Percent Purity 97.5%
Grade Pure
Physical Form Agglomerating Crystals
Chemical Name or Material p-Toluenesulfonic acid monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463016 Certificate TypeCertificate of Analysis Date17/05/2024 Catalog Number171785000,171780050,171780010,171780025
Lot NumberA0461316 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number171785000,171780050,171780010,171780025
Lot NumberA0460297 Certificate TypeCertificate of Analysis Date14/02/2024 Catalog Number171785000,171780050,171780010,171780025
Lot NumberA0444323 Certificate TypeCertificate of Analysis Date12/01/2024 Catalog Number171785000,171780050,171780010,171780025
Lot NumberA0423632 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog Number171785000,171780050,171780010,171780025
compliance-icons
Product Identifier
  • p-Toluenesulfonic acid monohydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.