Learn More
p-Thiocresol, 98%, Thermo Scientific Chemicals
Description
p-Thiocresol is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
p-Thiocresol is used as a pharmaceutical intermediate.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Air sensitive. Store away from oxidizing agents, air.

Specifications
Specifications
Melting Point | 40°C to 44°C |
Boiling Point | 195°C |
Flash Point | 68°C (154°F) |
Odor | Stench |
Quantity | 500 g |
UN Number | UN2811 |
Beilstein | 605761 |
Sensitivity | Air Sensitive |
Merck Index | 14,9325 |
Solubility Information | Soluble in alcohol. Insoluble in water. |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0400618 | Certificate of Analysis | 11/11/2022 | B24970.22,B24970.14,B24970.36 |
A0438478 | Certificate of Analysis | 11/11/2022 | B24970.22,B24970.14,B24970.36 |
10238203 | Certificate of Analysis | 12/04/2022 | B24970.22,B24970.14,B24970.36 |
10237908 | Certificate of Analysis | 12/04/2022 | B24970.22,B24970.14,B24970.36 |
10126953 | Certificate of Analysis | 23/11/2021 | B24970.22,B24970.14,B24970.36 |
Safety and Handling

- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.