missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Linezolid Antimicrobial Susceptibility discs

Determine the susceptibility of microorganisms using manual AST methods and a full range of high quality, easy-to-use antimicrobial susceptibility testing discs.

Brand:  Thermo Scientific™ CT1649B

43.46 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 11459848

  • 45.75€ / Pack of 250
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Linezolid Antimicrobial susceptibility discs.

Ease of use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.

disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results, but also provides a visual indication of the following:

  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics

Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Linezolid
10μg
LZD
Antimicrobial Susceptibility
50 Discs/Cartridge
Complies with BSAC, EUCAST guidelines
Oxoid Linezolid Antimicrobial Susceptibility Discs
Antimicrobial Susceptibility Discs
5 Cartridges/Pk.
50 Discs/Cartridge
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6224279 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog NumberCT1649B
Lot Number6224278 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog NumberCT1649B
Lot Number6199979 Certificate TypeCertificate of Analysis Date06/12/2024 Catalog NumberCT1649B
Lot Number6196777 Certificate TypeCertificate of Analysis Date24/11/2024 Catalog NumberCT1649B
Lot Number6187155 Certificate TypeCertificate of Analysis Date31/10/2024 Catalog NumberCT1649B
Promotions

Promotions

  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Oxoid™ Linezolid Antimicrobial Susceptibility discs >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.