Learn More
Thermo Scientific™ Oxoid™ CO2 Gen™ Sachet
Generate carbon dioxide atmosphere in AnaeroJar™ 2.5L with Oxoid CO2 Gen Sachet.
Brand: Thermo Scientific™ CD0025A
48.04 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Generate carbon dioxide atmosphere in AnaeroJar™ 2.5L with Oxoid CO2 Gen Sachet.
- No water or catalyst required
- No hydrogen gas produced
Specifications
CO2 Generating Sachet | |
Gas Generation Sachet | |
Thermo Scientific Oxoid | |
10/Pk. |
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%