Learn More
WTW™ OxiCal -ST™ Calibration and storage vessel
The calibration and storage containers for the WTW oxygen sensors are suitable for the standard conform calibration (DIN ISO 5814 or DIN ISO 17289: 2014-12) with water steam saturated air as well as for storage of the sensors to protect the sensitive membrane.
Brand:  WTW™ 205349
214.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10613782
  • 226.00€ / Each
Estimated Shipment: 13-06-2025
to see stock.
Explore more special offers
Product Title
WTW™ OxiCal -ST™ Calibration and storage vessel > Model: OxiCal -ST