missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid 0.25 M For Laboratory Use and Titrations, Chem Lab

Brand:  Chem Lab CL05.1503.1000


Product Code. 11963263

  • 67.05€ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

6153-56-6
100°C
0.25 M
1.01 g/mL
Polyethylene
Reference standards acc. NIST SRM 84l
0°C
Colorless
126.07
1 L
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.

Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.

Supplemental information
  • MIXTURE LIST-Contains : Oxalic acid

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Oxalic Acid 0.25 M For Laboratory Use and Titrations, Chem Lab > Oxalic Acid 0.25 M For Laboratory Use and Titrations, 1 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.