Learn More
BINDER™ Overheating Alarm Function for Drying and Heating Chambers
The Overheating Alarm Function for Drying and Heating Chambers features a switchable acoustic alarm, with adjustable limit value on the independent temperature safety device.
Brand:  BINDER™ 8012-1301
 View more versions of this product
Product Code. 16454169
  • 795.00€ / Each
Estimated Shipment: 04-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Audible alarm function for overheating
  • Features switchable acoustic alarm
  • Features adjustable limit value
  • Designed for use with BINDER™ Drying and Heating Chambers
Specifications
Specifications
Alarm function for overheating - Switchable acoustic alarm, with adjustable limit value on the independent temperature safety device
BINDER Chambers FED 400 Drying and Heating Chambers with Forced Convection and Enhanced Timer Functions, 400 V
Adjustable Overheating Alarm
400 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BINDER™ Overheating Alarm Function for Drying and Heating Chambers > For Use With: BINDER™ FED 400 Drying and Heating Chambers with Forced Convection and Enhanced Timer Functions, 400 V Model