missing translation for 'onlineSavingsMsg'
Learn More
Learn More
240 mV @ 25°C ORP Test Solutions, Hanna Instruments™
Click to view available options
Quantity:
230 mL
500 mL
Description
- Quality ORP test solution made from reagent grade chemicals and used to test platinum and gold ORP electrodes
- Designed for testing the performance of ORP electrodes
- Prepared with reagent grade chemicals
- Allows for a quick check of your ORP electrode
- Air tight bottle with tamper-proof seal to ensure quality
Specifications
Specifications
Quantity | 230 mL |
Odor | Odorless |
Physical Form | Liquid |
Concentration | 240mV |
Color | Yellow |
pH | 7 |
Solubility | Soluble |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
240 mV @ 25°C ORP Test Solutions, Hanna Instruments™ >
Spot an opportunity for improvement?Share a Content Correction