Learn More
Thermo Scientific™ Orion™ Probe Maintenance Kits for Dissolved Oxygen Meters
Help ensure optimal performance of dissolved oxygen probes with these specialized probe maintenance kits.
Brand:  Thermo Scientific™ 080513
 View more versions of this product
Product Code. 15942111
Description
Description
Help ensure optimal performance of Orion polarographic dissolved oxygen probes with Thermo Scientific™ Orion™ Probe Maintenance Kits for Dissolved Oxygen Meters, which include electrolyte solution, polishing disk and two membrane caps.
Warranty: Out of box.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For 083005MD, 083010MD, 083025MD, 083060MD, 086030MD, 083005A, 083010A, 083005D, 086020A, 081010F
Dissolved Oxygen Probe and Meter
Dissolved Oxygen Probe Accessories
2 Membrane caps, electrolyte solution, and polishing disk
Product Title
Thermo Scientific™ Orion™ Probe Maintenance Kits for Dissolved Oxygen Meters > Probe maintenance kit
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.