Learn More
Thermo Scientific™ Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe
Reduce probe servicing time and eliminate assembly problems using the Thermo Scientific™ Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe.
Brand:  Thermo Scientific™ 081003
 View more versions of this product
Product Code. 11767757
Description
Description
Reduce probe servicing time and eliminate assembly problems using Thermo Scientific™ Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe. Convenient screw-on membrane caps help ensure optimal probe performance.
 Warranty: Out of box
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Orion DO Probes 080510MD, 081010MD, 081010
Dissolved Oxygen Probe Accessories
Three Caps
Polarographic DO
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe > For use with 080510MD, 081010MD, 081010
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.