Learn More
Thermo Scientific™ Orion™ Ionplus™ Sure-Flow™ Cyanide Combination Ion Selective Electrode
Measure cyanide ions in aqueous solutions quickly, simply, accurately and economically.
504.56€ - 1045.20€
Specifications
Junction | Free-flowing liquid junction assures stable, drift-free potentials |
---|---|
For Use With (Equipment) | ISE Meters |
Product Code | Brand | Connector Types | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connector Types | Price | Quantity & Availability | ||||||
10039150
|
Thermo Scientific™
9406BN |
BNC |
1045.20€
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
10232591
|
Thermo Scientific™
9406SC |
Screw Cap | N/A | |||||||
10549131
|
Thermo Scientific™
9606BNWP |
Waterproof BNC |
887.42€
Each |
In Stock
Log in to see stock. |
||||||
Description
Measure cyanide ions in aqueous solutions quickly, simply and economically with Thermo Scientific™ Orion™ Cyanide Electrodes.
The ThermoFisher Orion Cyanide electrode complies with SM 4500-CN- F-1999 for Drinking water ; and SM 4500-CN- F-1999, ASTM D2036-09 (A)Wastewater measurements.
Features
- 9606BNWP is a combination electrode with a Sure-Flow™ junction. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.
Solutions required
- ISA
- 951011 ISA for cyanide measurements, 475 mL bottle
- Electrode fill solution
- 900062 Optimum Results B electrode filling solution, 5 x 60 mL bottles
Items included with
- 900062 Optimum Results B electrode filling solution. 60mL 1 bottle
- 948201 Polishing strip. Qty 2
Warranty period
- 12 months from date of purchase
Specifications
Free-flowing liquid junction assures stable, drift-free potentials | |
ISE Meters |