Learn More
Thermo Scientific™ Orion™ 2110XP Ammonia Analyzer
Control and optimize boiler ammonia levels with continuous, direct and precise ammonia measurements from the Thermo Scientific™ Orion™ 2110XP Ammonia Analyzer.
16570.00€
Specifications
Accuracy | ±15% or 30 ppb, whichever is greater |
---|---|
Accuracy (mV) | ± (0.5 mV + 0.1%) |
Accuracy (Temperature) | ±0.5°C |
Alarm Output | 3 |
Analog Outputs | 2, Galvanically Isolated |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
17151801
|
Thermo Scientific™
2110XPEN |
2110XP Ammonia Analyzer with Protective Enclosure |
16570.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reduce operational costs while producing the highest purity steam with the Thermo Scientific™ Orion™ 2110XP Ammonia Analyzer. This analyzer provides continuous, direct and precise measurements for the control and optimization of boiler ammonia levels.
Features
- Accurate detection allows precise ammonia control with confidence — maintain proper chemical feed levels without the guess work or excessive operating cost.
- Stable and drift-free measurements with minimal maintenance eliminate the need for frequent calibrations, saving you time and money.
- Repeatable and verifiable ammonia measurements using ammonia electrode technology for the highest level of performance available.
- Designed for ammonia specificity, the unique chemistry provides unsurpassed results across a wide range without the pitfalls of pH and conductivity “bottom out effects.”
- Step-by-step calibration process ensures a successful calibration the first time and every time.
- Ammonia detection range of 0 to 10ppm provides direct, accurate and precise results.
- Continuous online measurement ensures real-time information for optimum control of the ammonia feed without excessive operating costs.
- Selective and sensitive for ammonia, our system surpasses the performance of other systems that have the limitations of pH and conductivity to track real-time boiler ammonia levels.
2110XP Ammonia Analyzer Ordering Information
- Annual Consumables Kit, Cat. No. 211050XP
- Reagent, Cat. No. 181030
- Diffusion Tubing, Cat. No. 150060
- Calibration Kit, Cat. No. 2110XPCAL
- Calibration Standards, Cat. No. 181040
- Ammonia Electrode, Cat. No. 100047
- Reference Electrode, Cat. No. 210056
- Reference Electrode Filling Solution, Cat. No. 181073
Specifications
±15% or 30 ppb, whichever is greater | |
±0.5°C | |
2, Galvanically Isolated | |
Valox 364 | |
1 | |
2 | |
Ammonia Analyzer | |
Waterproof; IP66; NEMA 4X | |
2110XP ammonia analyzer with Ammonia electrode, reference electrode (100056), ATC probe (2100TP),ammonia calibration standards kit, CD user guide and options kit | |
Membrane | |
0 to 10ppm | |
0 to 20 mA; 4 to 20 mA | |
3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A | |
0.1°C | |
1/4 in. NPTF tube fitting | |
5 to 45°C | |
11 in. | |
17 in. | |
26 in. | |
85 to 132 VAC; 170 to 264 VAC | |
0 to 60 Hz | |
Optional ph/ORP or Conductivity |
± (0.5 mV + 0.1%) | |
3 | |
Double Known Addition | |
CE, CSA, UL, FCC Class A limits | |
Modbus with Additional Module | |
5°C to 35°C | |
Custom backlit LCD with temperature, concentration, mV, error codes and menu driven prompts | |
40mL/min. | |
8 to 100psig | |
5 to 95%, non-condensing | |
Wall mount | |
±1999.9m | |
0.1mV | |
95% within 2 minutes of injecting a standard solution | |
1 | |
11 x 17 x 26 in. (27 x 45 x 65cm) | |
27 cm | |
45 cm | |
65 cm | |
5A | |
Ammonia |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H361-Suspected of damaging fertility or the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
Your input is important to us. Please complete this form to provide feedback related to the content on this product.