Learn More
Ohaus™ OIML E2 Weight Set
Adjusted to Meet or Exceed OIML E2 Tolerances
462.00€ - 4395.00€
Product Code | Brand | Mass | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | ||||||
16682362
|
Ohaus™
30518028 |
500 mg to 1 mg |
462.00€
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||||
16692362
|
Ohaus™
30518029 |
50 g to 1 mg |
1200.00€
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||||
16662372
|
Ohaus™
30518036 |
50 g to 1 g |
800.00€
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||||
16602372
|
Ohaus™
30518030 |
100 g to 1 mg |
1325.00€
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||||
16672372
|
Ohaus™
30518037 |
100 g to 1 g |
908.00€
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||||
16612372
|
Ohaus™
30518031 |
2 x 200 g to 1 mg |
1610.00€
Each |
In Stock
Log in to see stock. |
||||||
16682372
|
Ohaus™
30518038 |
2 x 200 g to 1 g |
1210.00€
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||||
16622372
|
Ohaus™
30518032 |
500 g to 1 mg |
1860.00€
Each |
In Stock
Log in to see stock. |
||||||
16692372
|
Ohaus™
30518039 |
500 g to 1 g |
1475.00€
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||||
16632372
|
Ohaus™
30518033 |
1 kg to 1 mg |
2365.00€
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||||
16602382
|
Ohaus™
30518040 |
1 kg to 1 g |
1950.00€
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||||
16642372
|
Ohaus™
30518034 |
2 x 2 kg to 1 mg |
3550.00€
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||||
16612382
|
Ohaus™
30518041 |
2 x 2 kg to 1 g |
3155.00€
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||||
16652372
|
Ohaus™
30518035 |
5 kg to 1 mg |
4395.00€
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||||
16622382
|
Ohaus™
30518042 |
5 kg to 1 g |
3980.00€
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||||
Description
- Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
No Certificate | |
Protective Case |
Product Title
Ohaus™ OIML E2 Weight Set