Learn More
Implen OD600™ IQOQ Package Documentation
IQ/OQ Documentation package for the Implen OD600 (provided as Excel on USB stick) including two Neutral Density Glass filters.
Brand: Implen Implen OD600-Q
This item is not returnable.
View return policy
Description
- IQOQ package for validation of the Implen OD600 instrument.
- Includes
- Two glass filters for validation of photometric range and accuracy.
- IQOQ documentation provided as Excel file on USB stick
Specifications
Implen OD600 | |
IQ/OQ Documentation |
Product Content Correction
Product Title
Implen OD600™ IQOQ Package Documentation > IQOQ package