Learn More
Product Code | Brand | For Use With (Equipment) | Focus Distance | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Focus Distance | Price | Quantity & Availability | |||||
11917644
|
Vision Engineering™
MCO006 |
Mantis Compact Stereo Microscope | 68 mm |
430.96€
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
11947634
|
Vision Engineering™
MEO004 |
Mantis Elite Stereo Microscope | 96 mm |
322.36€
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
11907644
|
Vision Engineering™
MCO004 |
Mantis Compact Stereo Microscope | 96 mm |
247.26€
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
11977634
|
Vision Engineering™
MEO010 |
Mantis Elite Stereo Microscope | 54 mm |
1190.06€
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
11997634
|
Vision Engineering™
MEO020 |
Mantis Elite Stereo Microscope | 29 mm |
1917.96€
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
Product Title
Vision Engineering™ Objective Lens