Learn More
Brady™ Single Pole Circuit Breaker Lockout
Designed for 120 Volt breakers that have holes in the switch tongue.
Brand: Brady™ 065688
This item is not returnable.
View return policy
Description
- Quick and easy - just snap in place and apply lock
- Dual cavities accommodate different size switch tongues
Specifications
Single Pole Breaker Lockout | |
Nylon | |
54.50 x 15.20 mm | |
6 Pk. |
Red | |
UL 94 HB | |
15.2 mm | |
54.5 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Product Title
Brady™ Single Pole Circuit Breaker Lockout > Height: 15.2mm