Learn More
Socorex™ Nylon Cleaning Brushes
23.90€ - 44.30€
Specifications
Product Type | Cleaning Brush |
---|---|
Bristle Material | Nylon |
Handle Material | Plastic |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
10753643
|
Socorex™
1.170.300 |
Self-filling Syringe Set |
44.30€
1 set |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
10744633
|
Socorex™
1.170.301 |
0.3/0.5/1 mL Self-filling Syringes |
44.20€
Pack of 6 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
10559992
|
Socorex™
1.170.320 |
5/10/20 mL Self-filling Syringes |
23.90€
Pack of 3 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
10029931
|
Socorex™
1.170.305 |
1/2/5 mL Self-filling Syringes |
44.20€
Pack of 6 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
Specifications
Cleaning Brush | |
Nylon | |
Plastic |
Product Title
Socorex™ Nylon Cleaning Brushes